Assay Details
| Gene Symbol: MCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244177.1 | NP_001231106.1 | 3 - 4 | 762 | 86 | |
| GenBank mRNA | BC148891.1 | - | 2 - 3 | 642 | 86 | |
More Information
Set Membership: |
|

MCAT
Genomic Map