Assay Details
| Gene Symbol: RRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244181.1 | NP_001231110.1 | 7 - 8 | 850 | 66 | |
| NM_001244182.1 | NP_001231111.1 | 6 - 7 | 706 | 66 | ||
| GenBank mRNA | BC105511.1 | - | 6 - 7 | 647 | 66 | |
| DV889429.1 | - | 3 - 4 | 270 | 66 | ||
More Information
Set Membership: |
|

RRM2
Genomic Map