Assay Details
| Gene Symbol: LOC508666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244199.1 | NP_001231128.1 | 3 - 4 | 584 | 124 | |
| GenBank mRNA | BC110158.1 | - | 3 - 4 | 554 | 124 | |
| EV633346.1 | - | 3 - 4 | 587 | 124 | ||
More Information
Set Membership: |
|

LOC508666
Genomic Map