Assay Details
| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191348.3 | NP_001178277.1 | 2 - 3 | 827 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ORAI2| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191348.3 | NP_001178277.1 | 2 - 3 | 827 | 79 | |
Set Membership: |
|