Assay Details
| Gene Symbol: NR3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191349.2 | NP_001178278.1 | 4 - 5 | 2363 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR3C2| Gene Symbol: NR3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191349.2 | NP_001178278.1 | 4 - 5 | 2363 | 68 | |
Set Membership: |
|