Assay Details
| Gene Symbol: DALRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191397.1 | NP_001178326.1 | 3 - 4 | 725 | 62 | |
| GenBank mRNA | DN286402.1 | - | 3 - 4 | 699 | 62 | |
| DT815233.1 | - | 2 - 3 | 401 | 62 | ||
| EV635714.1 | - | 3 - 4 | 726 | 62 | ||
More Information
Set Membership: |
|

DALRD3
Genomic Map