Assay Details
| Gene Symbol: PACSIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191420.1 | NP_001178349.1 | 5 - 6 | 721 | 70 | |
| GenBank mRNA | DV931028.1 | - | 3 - 4 | 441 | 70 | |
More Information
Set Membership: |
|

PACSIN3
Genomic Map