Assay Details
| Gene Symbol: GSDMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244217.1 | NP_001231146.1 | 10 - 11 | 1237 | 78 | |
| GenBank mRNA | BC149305.1 | - | 9 - 10 | 3054 | 78 | |
| CK960776.1 | - | 3 - 4 | 233 | 78 | ||
More Information
Set Membership: |
|

GSDMB
Genomic Map