Assay Details
| Gene Symbol: GSDMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244217.1 | NP_001231146.1 | 3 - 4 | 491 | 95 | |
| GenBank mRNA | BC149305.1 | - | 3 - 4 | 492 | 95 | |
More Information
Set Membership: |
|

GSDMB
Genomic Map