Assay Details
| Gene Symbol: PANX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245925.1 | NP_001232854.1 | 3 - 4 | 551 | 67 | |
| GenBank mRNA | HM036582.1 | - | 3 - 4 | 551 | 67 | |
More Information
Set Membership: |
|

PANX1
Genomic Map