Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173959.4 | NP_776384.3 | 4 - 5 | 790 | 70 | |
| GenBank mRNA | AB075020.1 | - | 4 - 5 | 1033 | 70 | |
| AF188710.1 | - | 4 - 5 | 646 | 70 | ||
| AY241933.1 | - | 4 - 5 | 790 | 70 | ||
| BC112700.1 | - | 4 - 5 | 1040 | 70 | ||
More Information
Set Membership: |
|

SCD
Genomic Map