Assay Details
| Gene Symbol: SDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174178.2 | NP_776603.1 | 11 - 12 | 1567 | 67 | |
| GenBank mRNA | BC105357.1 | - | 11 - 12 | 1598 | 67 | |
| BT030722.1 | - | 11 - 12 | 1553 | 67 | ||
| DQ402990.1 | - | 9 - 10 | 1379 | 67 | ||
| M60879.1 | - | 11 - 12 | 1567 | 67 | ||
More Information
Set Membership: |
|

SDHA
Genomic Map