Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174246.2 | NP_776671.1 | 3 - 4 | 982 | 118 | |
| GenBank mRNA | X83741.1 | - | 982 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AVPR2| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174246.2 | NP_776671.1 | 3 - 4 | 982 | 118 | |
| GenBank mRNA | X83741.1 | - | 982 | 118 | ||
Set Membership: |
|