Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174518.1 | NP_776943.1 | 1 - 2 | 132 | 69 | |
| GenBank mRNA | AF362075.1 | - | 1 - 2 | 132 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174518.1 transcripts.
Set Membership: |
|

CITED1
Genomic Map