Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174518.1 | NP_776943.1 | 2 - 3 | 250 | 84 | |
| GenBank mRNA | AF362075.1 | - | 2 - 3 | 250 | 84 | |
| BC102725.1 | - | 2 - 3 | 217 | 84 | ||
More Information
Set Membership: |
|

CITED1
Genomic Map