Assay Details
| Gene Symbol: IGFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174554.3 | NP_776979.2 | 2 - 3 | 637 | 75 | |
| GenBank mRNA | BC111197.1 | - | 2 - 3 | 638 | 75 | |
| X54979.1 | - | 629 | 75 | |||
More Information
Set Membership: |
|

IGFBP1
Genomic Map