Assay Details
| Gene Symbol: IGFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174554.3 | NP_776979.2 | 3 - 4 | 766 | 93 | |
| GenBank mRNA | BC111197.1 | - | 3 - 4 | 767 | 93 | |
| X54979.1 | - | 758 | 93 | |||
More Information
Set Membership: |
|

IGFBP1
Genomic Map