Assay Details
| Gene Symbol: ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177493.2 | NP_803459.1 | 7 - 8 | 946 | 63 | |
| GenBank mRNA | J02671.1 | - | 7 - 8 | 946 | 63 | |
| M81862.1 | - | 7 - 8 | 949 | 63 | ||
More Information
Set Membership: |
|

ASMT
Genomic Map