Assay Details
| Gene Symbol: RET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191483.1 | NP_001178412.1 | 17 - 18 | 3135 | 57 | |
| GenBank mRNA | BC149315.1 | - | 17 - 18 | 3136 | 57 | |
More Information
Set Membership: |
|

RET
Genomic Map