Assay Details
| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191533.1 | NP_001178462.1 | 7 - 8 | 855 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2R2A| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191533.1 | NP_001178462.1 | 7 - 8 | 855 | 63 | |
Set Membership: |
|