Assay Details
| Gene Symbol: HNRNPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192030.2 | NP_001178959.1 | 10 - 11 | 1565 | 61 | |
| GenBank mRNA | BC108092.1 | - | 10 - 11 | 1525 | 61 | |
More Information
Set Membership: |
|

HNRNPL
Genomic Map