Assay Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192041.1 | NP_001178970.1 | 7 - 8 | 706 | 86 | |
| GenBank mRNA | BC105269.1 | - | 7 - 8 | 676 | 86 | |
More Information
Set Membership: |
|

DOLPP1
Genomic Map