Assay Details
Gene Symbol: EHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192053.1 | NP_001178982.1 | 1 - 2 | 322 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
