Assay Details
| Gene Symbol: POLR2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192065.2 | NP_001178994.1 | 1 - 2 | 1260 | 84 | |
| GenBank mRNA | BC118247.1 | - | 1 - 2 | 93 | 84 | |
More Information
Set Membership: |
|

POLR2H
Genomic Map