Assay Details
| Gene Symbol: MYBPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192119.1 | NP_001179048.1 | 7 - 8 | 1058 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBPHL| Gene Symbol: MYBPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192119.1 | NP_001179048.1 | 7 - 8 | 1058 | 62 | |
Set Membership: |
|