Assay Details
| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192149.1 | NP_001179078.1 | 3 - 4 | 278 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF9B| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192149.1 | NP_001179078.1 | 3 - 4 | 278 | 100 | |
Set Membership: |
|