Assay Details
| Gene Symbol: C1RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001354760.1 | NP_001341689.1 | 5 - 6 | 749 | 85 | |
| GenBank mRNA | EV615074.1 | - | 4 - 5 | 468 | 85 | |
More Information
Set Membership: |
|

C1RL
Genomic Map