Assay Details
| Gene Symbol: CA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192205.1 | NP_001179134.1 | 4 - 5 | 1224 | 73 | |
| GenBank mRNA | BC103148.1 | - | 4 - 5 | 686 | 73 | |
More Information
Set Membership: |
|

CA14
Genomic Map