Assay Details
| Gene Symbol: IMPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192282.1 | NP_001179211.1 | 4 - 5 | 573 | 65 | |
| GenBank mRNA | BC153859.1 | - | 2637 | 65 | ||
More Information
Set Membership: |
|

IMPA2
Genomic Map