Assay Details
| Gene Symbol: GNB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192340.2 | NP_001179269.1 | 4 - 5 | 578 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNB1L| Gene Symbol: GNB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192340.2 | NP_001179269.1 | 4 - 5 | 578 | 87 | |
Set Membership: |
|