Assay Details
| Gene Symbol: ZNF428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192364.1 | NP_001179293.1 | 1 - 2 | 65 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192364.1 transcripts.
Set Membership: |
|

ZNF428
Genomic Map