Assay Details
| Gene Symbol: STARD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192379.1 | NP_001179308.1 | 1 - 2 | 100 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192379.1 transcripts.
Set Membership: |
|

STARD10
Genomic Map