Assay Details
| Gene Symbol: MARCHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192387.2 | NP_001179316.2 | 2 - 3 | 212 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCHF10| Gene Symbol: MARCHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192387.2 | NP_001179316.2 | 2 - 3 | 212 | 89 | |
Set Membership: |
|