Assay Details
| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192389.1 | NP_001179318.1 | 6 - 7 | 1596 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX59| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192389.1 | NP_001179318.1 | 6 - 7 | 1596 | 79 | |
Set Membership: |
|