Assay Details
| Gene Symbol: CA5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192409.1 | NP_001179338.1 | 3 - 4 | 543 | 65 | |
| GenBank mRNA | CB441532.1 | - | 3 - 4 | 543 | 65 | |
| CB441881.1 | - | 4 - 5 | 573 | 65 | ||
More Information
Set Membership: |
|

CA5A
Genomic Map