Assay Details
| Gene Symbol: CA5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192409.1 | NP_001179338.1 | 4 - 5 | 638 | 98 | |
| GenBank mRNA | CB441881.1 | - | 3 - 4 | 478 | 98 | |
More Information
Set Membership: |
|

CA5A
Genomic Map