Assay Details
| Gene Symbol: FABP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192410.1 | NP_001179339.1 | 3 - 4 | 365 | 90 | |
| GenBank mRNA | CF768059.1 | - | 3 - 4 | 365 | 90 | |
More Information
Set Membership: |
|

FABP9
Genomic Map