Assay Details
| Gene Symbol: CEMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192415.1 | NP_001179344.1 | 13 - 14 | 2549 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEMIP2| Gene Symbol: CEMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192415.1 | NP_001179344.1 | 13 - 14 | 2549 | 69 | |
Set Membership: |
|