Assay Details
| Gene Symbol: TJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192419.1 | NP_001179348.1 | 5 - 6 | 502 | 68 | |
| GenBank mRNA | BC105505.1 | - | 5 - 6 | 487 | 68 | |
More Information
Set Membership: |
|

TJAP1
Genomic Map