Assay Details
| Gene Symbol: DGKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192930.3 | NP_001179859.3 | 1 - 2 | 456 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGKE| Gene Symbol: DGKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192930.3 | NP_001179859.3 | 1 - 2 | 456 | 72 | |
Set Membership: |
|