Assay Details
| Gene Symbol: TFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192976.2 | NP_001179905.1 | 6 - 7 | 1033 | 60 | |
| GenBank mRNA | BC104525.2 | - | 6 - 7 | 969 | 60 | |
| EH198200.1 | - | 3 - 4 | 367 | 60 | ||
More Information
Set Membership: |
|

TFG
Genomic Map