Assay Details
| Gene Symbol: PCYT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193051.1 | NP_001179980.1 | 5 - 6 | 565 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCYT1B| Gene Symbol: PCYT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193051.1 | NP_001179980.1 | 5 - 6 | 565 | 73 | |
Set Membership: |
|