Assay Details
| Gene Symbol: TCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193105.2 | NP_001180034.1 | 8 - 9 | 1236 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCN1| Gene Symbol: TCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193105.2 | NP_001180034.1 | 8 - 9 | 1236 | 89 | |
Set Membership: |
|