Assay Details
| Gene Symbol: MARCHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193149.1 | NP_001180078.1 | 2 - 3 | 403 | 75 | |
| GenBank mRNA | EV608718.1 | - | 2 - 3 | 412 | 75 | |
More Information
Set Membership: |
|

MARCHF1
Genomic Map