Assay Details
| Gene Symbol: SCNN1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193174.2 | NP_001180103.1 | 12 - 13 | 1942 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCNN1G| Gene Symbol: SCNN1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193174.2 | NP_001180103.1 | 12 - 13 | 1942 | 54 | |
Set Membership: |
|