Assay Details
| Gene Symbol: EPHX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193176.1 | NP_001180105.1 | 5 - 6 | 806 | 70 | |
| GenBank mRNA | BC151697.1 | - | 5 - 6 | 1031 | 70 | |
More Information
Set Membership: |
|

EPHX3
Genomic Map