Assay Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195018.1 | NP_001181947.1 | 1 - 2 | 197 | 79 | |
| GenBank mRNA | BC149331.1 | - | 1 - 2 | 198 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149331.1, NM_001195018.1 transcripts.
Set Membership: |
|

ZMYM5
Genomic Map