Assay Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195018.1 | NP_001181947.1 | 3 - 4 | 775 | 84 | |
| GenBank mRNA | BC149331.1 | - | 3 - 4 | 776 | 84 | |
More Information
Set Membership: |
|

ZMYM5
Genomic Map