Assay Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195018.1 | NP_001181947.1 | 5 - 6 | 1147 | 89 | |
| GenBank mRNA | BC149331.1 | - | 5 - 6 | 1148 | 89 | |
More Information
Set Membership: |
|

ZMYM5
Genomic Map