Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195631.1 | NP_001182560.1 | 7 - 8 | 798 | 94 | |
| GenBank mRNA | BC149051.1 | - | 5 - 6 | 1107 | 94 | |
More Information
Set Membership: |
|

CCDC189
Genomic Map